[beta-Ala8]Neurokinin A (4-10)
Supplier: ENZO LIFE SCIENCES
BMLP5060001
ENZOBMLP5060001EA 161 EUR
ENZOBMLP5060001
[beta-Ala8]Neurokinin A (4-10)
Proteins and Peptides
Neurokinin receptor ligand
Highly selective NK2 neurokinin receptor agonist.