Dihydrosphingosine-1-phosphate, D-erythro ≥98%

Supplier: ENZO LIFE SCIENCES

BMLSL1430001
ENZOBMLSL1430001EA 211 EUR
ENZOBMLSL1430001
Dihydrosphingosine-1-phosphate, D-erythro ≥98%
Dihydrosphingosine-1-phosphate, D-erythro

S1P receptor ligand.


Saturated analog of sphingosine-1-phosphate. Induces chemotaxis in HUVECs. May be used as a negative control for intracellular effects of S1P. Rapidly dephosphorylated by a sphingosine phosphatase in cultured human fibroblasts. Ligand for S1P (EDG) receptors.


Soluble in methanol/THF/water (60/30/10) (0.5 mg/ml). Slightly soluble in DMSO, methanol or acetic acid.

Formula: C₁₈H₄₀NO₅P
MW: 381.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00871363
CAS Number: 28508-07-8

Order Now

Specification Test Results

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR