Nuclear-ID™ red/green cell viability reagent
Supplier: ENZO LIFE SCIENCES
ENZ53006C100
ENZOENZ53006C100EA 361 EUR
ENZOENZ53006C100
Nuclear-ID™ red/green cell viability reagent
Protein Linking and Labeling Systems
Cell viability dye
The Nuclear-ID™ Red/Green cell viability reagent (Prod. No. ENZ-53006) is a mixture of a red fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers.
Informace pro objednávání: Quantity: 100 μl (for 1000 microscopy assays).