Anti-ACTA2 Mouse Monoclonal Antibody

Supplier: ENZO LIFE SCIENCES
ENZC34933
ENZOENZC34933EA 361 EUR
ENZOENZC34933
Anti-ACTA2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunohistochemistry, Western Blot

Recognizes human, monkey, chicken and rabbit actin isotypes alpha and gamma of smooth muscle cells. Does not cross-react with actin of skeletal or cardiac muscle cells.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol ACTA2
Antigen name Smooth Muscle Actin
Environmentally Preferable
Antigen synonyms AAT6|alpha 2 actin|MYMY5|aortic smooth muscle|cell growth-inhibiting gene 46 protein|alpha-cardiac actin|alpha-actin-2|actin|GIG46|growth-inhibiting gene 46|ACTSA
Antibody type Primary
Clonality Monoclonal
Reactivity Human
Host Mouse
Isotype IgG2b
Western blot Yes
ImmunoChemistry Yes
Form Liquid. In PBS containing 0,1% sodium azide and 1% BSA.
Purification Affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR