Human recombinant Renin (active)

Supplier: ENZO LIFE SCIENCES

ENZPRT1930010
ENZOENZPRT1930010EA 245 EUR
ENZOENZPRT1930010
Human recombinant Renin (active)
Proteins and Peptides

Highly specific active enzyme cleaves angiotensinogen to yield angiotensin I


Produced in HEK cells. Mature Renin (aa 67-406) is fused at the N-terminus to a FLAG®-tag. Active form, does not contain prorenin peptide (aa 1-66).

New Product

Order Now

SPECIFICATIONS

Protein/peptide name Renin (active)
Environmentally Preferable
Protein synonyms angiotensinogen|Angiotensinogenase|Angiotensin forming enzyme|Ren
Protein/peptide type Recombinant
Species Human
Source
Purity >90%
Concentration 0,1 mg/ml when reconstituted as suggested.
Formulation Lyophilized. In 25 mM MES buffer containing 150 mM NaCl.
Molecular weight ~52 kDa (predicted is 38,3 kDa)
Purification Affinity purifed.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR