Water sampling bottles, glass, narrow neck, Boston type, round
Supplier: EP SCIENTIFIC
329-0500 329-1000 349-0500 329-0250 349-0125 129-0125 129-0250 129-0500 129-1000 229-0125 229-0250 229-0500 229-1000 329-0125
EPSC329-0500EA 101 EUR
EPSC329-0500 EPSC329-1000 EPSC349-0500 EPSC329-0250 EPSC349-0125 EPSC129-0125 EPSC129-0250 EPSC129-0500 EPSC129-1000 EPSC229-0125 EPSC229-0250 EPSC229-0500 EPSC229-1000 EPSC329-0125
Water sampling bottles, glass, narrow neck, Boston type, round
Bottles Water Sampling Bottles
These bottles, made of Type III soda-lime glas, clear or amber, with cap, are recommended for collecting water samples for variety of organic and inorganic target analytes where light protection is not required.
  • With PTFE-lined white PP closed top cap (assembled on bottles)
  • Certified to meet EPA performance based specifications for organics, pesticides and metal anlyses or unprocessed and readily assembled for own cleaning procedure
  • Each jar individually barcoded with lot number and unique container number
Delivery information: Supplied withMore Product Information