Sample vials, economy processed
Supplier: EP SCIENTIFIC
SB36-0040 SB46-0040 TB36-0040
EPSCSB36-0040EA 515 EUR
EPSCSB36-0040 EPSCSB46-0040 EPSCTB36-0040
Sample vials, economy processed
Vials
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Open-top white PP cap with choice of standard 3,18 mm or thinner 1,52 mm PTFE-lined silicone septum
  • Directly compatible with automation; thin septa vial is optimised for easy needle puncture
  • Vial shoulder slope optimised to prevent air bubble entrapment
  • With unattached sample analysis labels for optional use
Processed to meet or exceed EPA performance based specifications for volatile organic analysis but withoutMore Product Information