Restriction enzymes, EcoRI, Fermentas

Supplier: Thermo Fisher Scientific

ER0271 ER0272 ER0275
ER0271EA 49.4 EUR
ER0271 ER0272 ER0275
Restriction enzymes, EcoRI, Fermentas
Nucleic Acid Reagents Restriction Enzymes

These EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37 °C in its own unique buffer. Exhibit 100% activity in the recommended buffer and reaction conditions.


  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient color-coded five buffer system and BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities


Restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that sent in DNA preparations. Includes universal Tango buffer for double-digestions.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name EcoRI EcoRI
Source
Overhang type 3' overhang 5' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless) FastDigest Green Buffer or FastDigest Buffer (colorless)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR