Kit Mutation Generation (MGS) generation of random insertions 10 reactions. Kit includes MuA Transposase (10l), Entranceposon (M1-C amR) (10l), Entranceposon (M1-KanR) (10l), 5x reaction buffer for MuA (100l), control target DNA (10l), NotI Mini 1 * 1 KIT
Supplier: FINNZYMES REAGENTS
Kit Mutation Generation (MGS) generation of random insertions 10 reactions. Kit includes MuA Transposase (10l), Entranceposon (M1-C amR) (10l), Entranceposon (M1-KanR) (10l), 5x reaction buffer for MuA (100l), control target DNA (10l), NotI Mini 1 * 1 KIT