Dry Media Bag  with 15 bags and 2 filters. Demi-Fraser or Half Frasier Media is a modification of Fraser Broth Base, this media was developed by Fraser and Sperber for the rapid detection of Listeria from food and environmental samples. The addition 1 * 15 items
Supplier: FOODCHEK SYSTEMS
Dry Media Bag  with 15 bags and 2 filters. Demi-Fraser or Half Frasier Media is a modification of Fraser Broth Base, this media was developed by Fraser and Sperber for the rapid detection of Listeria from food and environmental samples. The addition 1 * 15 items