Transparent Tamper-Evident Cryogenic Tape for Frozen Containers
Supplier: GA
TEQ-22C3-100
GAINTEQ-22C3-100EA 90.5 EUR
GAINTEQ-22C3-100
Transparent Tamper-Evident Cryogenic Tape for Frozen Containers
Labels
Transparent tamper-evident tape for already frozen containers and boxes, ideal for long-term storage in cryogenic and deep-freeze conditions. Suitable for immersion and thawing in a water baths, the tape will tear upon any attempt to remove it, indicating any attempt to open the labeled container.
  • Temperature range: –196 to +121 °C (–321 to +250 °F)
  • Clear tamper-evident design indicates any attempt to open the container
  • Writable with permanent cryogenic markers
  • Ideal for already frozen surfaces (as low as –80 °C) without thawing first
Transparent tapeMore Product Information