Milk Butyrometer Original Gerber 0 - 10 % : 0.2, 10's pack Frosted label on bulb, plain neck, without stopper, for quantities of milk from 10.73 - 11 ml. Manufactured according to ISO standard 488 IDF 105. 1 * 10 items
Supplier: Gerber Instruments AG
Milk Butyrometer Original Gerber 0 - 10 % : 0.2, 10's pack Frosted label on bulb, plain neck, without stopper, for quantities of milk from 10.73 - 11 ml. Manufactured according to ISO standard 488 IDF 105. 1 * 10 items