Desalting column, G-Trap™ GT
Supplier: G-Biosciences
786-1025 786-1026 786-1027 786-1028 786-1023 786-1024
GENO786-1025EA 157 EUR
GENO786-1025 GENO786-1026 GENO786-1027 GENO786-1028 GENO786-1023 GENO786-1024
Desalting column, G-Trap™ GT
Chromatography Columns
G-Trap™ GT desalting columns are prepacked ready to use columns for separation of low molecular weight substances from high molecular weight substances based on size exclusion chromatography. Mostly used for removal of salt or buffer exchange before or after different chromatographic steps. They can also be used for separation of proteins based on their sizes.
  • Made of biocompatible PP, which does not interact with biomolecules
  • Stopper at the inlet and snap-off end at the outlet
The G-Trap™ GT desalting columns can be operated with syringe, peristalticMore Product Information