Ceaprene stopper
Supplier: Greiner Bio-One
330070_605
GREI330070_605EA 75.3 EUR
GREI330070_605
Ceaprene stopper
Stoppers Rubber Stoppers
This ceaprene stopper is made of water-repellent material.
  • Gas-permeable
  • Non-sterile
Used for closure of Drosophila containers.