Safety Blood Collection Set + Luer Adapter
Supplier: Greiner Bio-One
450081V1
GREI450081V1EA 754 EUR
GREI450081V1
Safety Blood Collection Set + Luer Adapter
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Safety blood collection set + luer adapter, single-packed, sterile.
  • Winged grip style
  • Safety needle, thin wall type
Safety feature: Retractable needle
Tubing length: 190,5 mm