Glass bottom microplates, SensoPlate™
Supplier: Greiner Bio-One
662892_12 655896_16 655892_16 781856_16 781892_16 783892_16 655891_16
738-0031EA 712 EUR
738-0031 GREI655896_16 738-0027 738-0504 738-0104 738-0105 GREI655891_16
Glass bottom microplates, SensoPlate™
Microplates HTS Microplates
Optically clear borosilicate glass bottom (light path 175±15 µm) and black PS frame.
  • All well formats available: 24, 96, 384 and 1536 wells
  • Standard SBS footprint
  • SensoPlate™ Plus option for high throughput screening applications
  • Free of DNase, RNase and human DNA, and non-pyrogenic
SensoPlate™ glass bottom microplates are designed for fluorescence correlation spectroscopy. The glass bottom allows transmission measurements in the wavelength range above 350 nm. Microscopic applications, such as confocal microscopy, are another potential area ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)