Cell culture dishes, cell-repellent surface, CELLSTAR®
Supplier: Greiner Bio-One
664970_5 627979_40
GREI664970_5EA 215 EUR
GREI664970_5 GREI627979_40
Cell culture dishes, cell-repellent surface, CELLSTAR®
Culture Dishes Cell Culture Dishes
These sterile, PS cell culture dishes have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • Lid with condensation rings
  • Other formats available (flasks, microplates, multiwell plates)
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)