Rack for cryotubes
Supplier: Greiner Bio-One
802501_5
GREI802501_5EA 248 EUR
GREI802501_5
Rack for cryotubes
Racks Cryogenic Vial Racks
PC, blue.
  • Non slip rubber base
This rack is suitable for cryotubes with a 'starfoot' base and allows one-handed manipulation of the tubes.