6-Fluoro-m-toluonitrile 99

Supplier: Thermo Fisher Scientific
Danger

H26166.03 H26166.06
H26166.03EA 18.1 EUR
H26166.03 H26166.06
6-Fluoro-m-toluonitrile 99
6-Fluoro-m-toluonitrile

Purity: 99% CAS No.: 64113-84-4 Molecular Formula: C8H6FN Formula Weight: 135.14 Flash Point: 94°(201°F) Melting Point: 49-52° UN No.: 3439 MDL No.: MFCD00134542

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 49…52 °C
Flash Pt: 94 °C (201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00134542
CAS Number: 64113-84-4
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR