2-Fluoro-4-hydroxybenzaldehyde 97

Supplier: Thermo Fisher Scientific
Warning

H26218.MCEA 105 EUR
H26218.MC H26218.ME
2-Fluoro-4-hydroxybenzaldehyde 97
2-Fluoro-4-hydroxybenzaldehyde

Purity: 97% CAS No.: 348-27-6 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 168-170° MDL No.: MFCD06797918

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06797918
CAS Number: 348-27-6
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR