Cyclopentyl methyl ether (CPME) ≥99.9%

Supplier: Thermo Fisher Scientific
Danger

H27349.AP H27349.AU H27349.0F H27349.50
H27349.APEA 69.8 EUR
H27349.AP H27349.AU H27349.0F H27349.50
Cyclopentyl methyl ether (CPME) ≥99.9%
Cyclopentyl methyl ether (CPME)

Purity: 99.9+% CAS No.: 5614-37-9 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: -1°(-30°F) Refractive Index: 1.4210 Melting Point: -140° Boiling Point: 106° UN No.: 3271 MDL No.: MFCD08276401

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 106 °C
Melting Pt: –140 °C
Density: 0.86
Flash Pt: –1 °C (–30 °F)
Storage Temperature: Ambient
MDL Number: MFCD08276401
CAS Number: 5614-37-9
UN: 3271
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR