3-Bromopyridine-2-carbonitrile 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-Bromopicolinonitrile, 3-Bromo-2-cyanopyridine

H27632.03 H27632.06
H27632.03EA 89 EUR
H27632.03 H27632.06
3-Bromopyridine-2-carbonitrile 98
3-Bromopyridine-2-carbonitrile

Purity: 98% CAS No.: 55758-02-6 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Melting Point: 96-99° Boiling Point: 120°/4mm MDL No.: MFCD02683288

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Boiling Pt: 120 °C (4 mmHg)
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD02683288
CAS Number: 55758-02-6
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR