4-Cyanophenylboronic acid pinacol ester 97

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester

H28208.06 H28208.03
H28208.06EA 205 EUR
H28208.06 H28208.03
4-Cyanophenylboronic acid pinacol ester 97
4-Cyanophenylboronic acid pinacol ester

Purity: 97% CAS No.: 171364-82-2 Molecular Formula: C13H16BNO2 Formula Weight: 229.08 MDL No.: MFCD03093897

Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093897
CAS Number: 171364-82-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR