N-Fmoc-D-leucine ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Fmoc-D-Leu-OH

H29041.06 H29041.14 H29041.03
H29041.06EA 91.4 EUR
H29041.06 H29041.14 H29041.03
N-Fmoc-D-leucine ≥98%
N-Fmoc-D-leucine

Purity: 98% CAS No.: 114360-54-2 Molecular Formula: C21H23NO4 Formula Weight: 353.42 Storage Temperature: 2°C to 8°C Melting Point: 151-154° Optical Rotation: +25° (c=1 in DMF) MDL No.: MFCD00062957

Formula: C₂₁H₂₃NO₄
MW: 353.4 g/mol
Melting Pt: 151…154 °C
MDL Number: MFCD00062957
CAS Number: 114360-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR