6-Morpholino-3-pyridinamine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine

H50699.03 H50699.MD H50699.06
H50699.03EA 217 EUR
H50699.03 H50699.MD H50699.06
6-Morpholino-3-pyridinamine 97%
6-Morpholino-3-pyridinamine

Purity: 97% CAS No.: 52023-68-4 Molecular Formula: C9H13N3O Formula Weight: 179.22 Melting Point: 127-130° MDL No.: MFCD00119562

Formula: C₉H₁₃N₃O
MW: 179.22 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00119562
CAS Number: 52023-68-4
EINECS: 257-613-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR