4-Chloro-6-quinolineboronic acid pinacol ester 96%

Supplier: Thermo Fisher Scientific

H54085.MD H54085.03
H54085.MDEA 104 EUR
H54085.MD H54085.03
4-Chloro-6-quinolineboronic acid pinacol ester 96%
4-Chloro-6-quinolineboronic acid pinacol ester

Purity: 96% Molecular Formula: C15H17BClNO2 Formula Weight: 289.56 MDL No.: MFCD16987803

Formula: C₁₅H₁₇BClNO₂
MW: 289.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987803
CAS Number: 1201844-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR