1,3-Difluoro-2-nitrobenzene 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,6-Difluoronitrobenzene

H55694.03 H55694.06
H55694.03EA 87.5 EUR
H55694.03 H55694.06
1,3-Difluoro-2-nitrobenzene 98
1,3-Difluoro-2-nitrobenzene

Purity: 98% CAS No.: 19064-24-5 Molecular Formula: C6H3F2NO2 Formula Weight: 159.09 Flash Point: 88°(190°F) Refractive Index: 1.4940 Melting Point: 0° Boiling Point: 91-92°/11mm MDL No.: MFCD00192035

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 91…92 °C (11 mmHg)
Density: 1.503
Flash Pt: 88 °C (190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00192035
CAS Number: 19064-24-5
EINECS: 242-793-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR