N-Allylaniline 95%

Supplier: Thermo Fisher Scientific
Danger

H55821.03 H55821.06 H55821.14
H55821.03EA 0 EUR
H55821.03 H55821.06 H55821.14
N-Allylaniline 95%
N-Allylaniline
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 218…220 °C
Density: 0.982
Flash Pt: 89 °C (192 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008638
CAS Number: 589-09-3
EINECS: 209-633-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR