Oxetane-3-carboxylic acid 95%

Supplier: Thermo Fisher Scientific
Warning

H57578.MDEA 460 EUR
H57578.MD H57578.ME
Oxetane-3-carboxylic acid 95%
Oxetane-3-carboxylic acid

Purity: 95% CAS No.: 114012-41-8 Molecular Formula: C4H6O3 Formula Weight: 102.09 Storage Temperature: -30°C to -10°C Flash Point: 115°(239°F) Boiling Point: 244° MDL No.: MFCD08669505

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 244 °C
Density: 1.374
Flash Pt: 115 °C (239 °F)
MDL Number: MFCD08669505
CAS Number: 114012-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR