2-Amino-4-(2-pyridyl)thiazole 97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(Pyridin-2-yl)thiazol-2-amine

H58554.04
H58554.04EA 302 EUR
H58554.04
2-Amino-4-(2-pyridyl)thiazole 97%
2-Amino-4-(2-pyridyl)thiazole

Purity: 97% CAS No.: 30235-26-8 Molecular Formula: C8H7N3S Formula Weight: 177.22 Physical Form: Solid Appearance: Pale brown MDL No.: MFCD00460417

Formula: C₈H₇N₃S
MW: 177.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00460417
CAS Number: 30235-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR