(R)-2-Aminopropanamide hydrochloride 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: H-D-Ala-NH2.HCl, H-D-Ala-NH₂·HCl, D-Alaninamide hydrochloride

H63535.06
H63535.06EA 184 EUR
H63535.06
(R)-2-Aminopropanamide hydrochloride 98%
(R)-2-Aminopropanamide hydrochloride

Purity: 98% CAS No.: 71810-97-4 Molecular Formula: C3H8N2O.HCl Formula Weight: 124.57 Physical Form: Powder Appearance: White MDL No.: MFCD00039093

Formula: C₃H₉ClN₂O
MW: 124.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039093
CAS Number: 71810-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR