Tetrahydropyran-4-carboxylic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tetrahydropyran-4-yl-carboxylic acid

H64340.06 H64340.14
H64340.06EA 60.4 EUR
H64340.06 H64340.14
Tetrahydropyran-4-carboxylic acid 98%
Tetrahydropyran-4-carboxylic acid

Purity: 98% CAS No.: 5337-03-1 Molecular Formula: C6H10O3 Formula Weight: 130.14 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: White to off-white Melting Point: 87-89° Beilstein Reference: 112249 MDL No.: MFCD00031016

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD00031016
CAS Number: 5337-03-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR