1-Aminocyclobutanecarboxylic acid 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Aminocyclobutane-1-carboxylic acid, 1-Amino-1-cyclobutanecarboxylic acid

H64459.03 H64459.06
H64459.03EA 60.3 EUR
H64459.03 H64459.06
1-Aminocyclobutanecarboxylic acid 95%
1-Aminocyclobutanecarboxylic acid

Purity: 95% CAS No.: 22264-50-2 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Physical Form: Powder Appearance: White Melting Point: 261° dec. Beilstein Reference: 2802253 MDL No.: MFCD00661068

Formula: C₅H₉NO₂
MW: 115.132 g/mol
Melting Pt: 261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00661068
CAS Number: 22264-50-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR