4-Bromo-1H-pyrazole 98+%

Supplier: Thermo Fisher Scientific
Warning

H64673.14 H64673.36
H64673.14EA 65 EUR
H64673.14 H64673.36
4-Bromo-1H-pyrazole 98+%
4-Bromo-1H-pyrazole

Purity: 98+% CAS No.: 2075-45-8 Molecular Formula: C3H3BrN2 Formula Weight: 146.97 Physical Form: Solid Melting Point: 93-96° lit. Boiling Point: 250-260° lit. Beilstein Reference: 106510 MDL No.: MFCD00075602

Formula: C₃H₃BrN₂
MW: 146.97 g/mol
Boiling Pt: 250…260 °C (lit.)
Melting Pt: 93…96 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00075602
CAS Number: 2075-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR