Test kit, chlorine and pH, model CN-67
Supplier: Hach
1411100
HACH1411100EA 324 EUR
HACH1411100
Test kit, chlorine and pH, model CN-67
Environmental Test Kits
Maintaining safe water conditions and to protect against system corrosion by control of chlorine and pH. The free and total chlorine and pH test kit provides more accuracy than test kits using colour cubes or step-wise comparators.
Delivery information: Includes colour discs and a comparator for monitoring chlorine and pH and required reagents and apparatus in a rugged carrying case for analysis in the field.