Multi parameter test kit
Supplier: Hach
223002
HACH223002EA 523 EUR
HACH223002
Multi parameter test kit
Environmental Test Kits
This multi-test kit combines popular water treatment tests into one compact kit which can be used to determine chlorine (0 to 3,4 mg/L), hardness (1 to 20 gpg), iron (0 to 4 mg/L) and pH (4 to 10) in the water sample.
  • Includes frequently measured water quality parameters
  • Uses simple, economical methods
Hardeness testing: gpg= grains per gallon
Delivery information: Includes six reagents, bottles, colour discs, comparator box, viewing tubes, measuring tube, instructions and carrying case.