Acidity reagent set for digital titrator
Supplier: Hach
2272800
HACH2272800EA 149 EUR
HACH2272800
Acidity reagent set for digital titrator
Environmental Test Kits
This set is used for the determination of acidity by digital titrator titration with methyl orange and phenolpthalein end points.
  • Fast and Flexible
  • Versatile
  • Economical
  • Accurate
Reagent Set includes: Bromphenol blue powder pillows, phenolphthalein powder pillows and two NaOH cartridges (0.1600 N and 1.600N). Ranges: 10 to 160 and 1000 to 4000 mg/L as CaCO₃.