Sodium thiosulphate 0.0246 mol/l (0.0246 N) in aqueous solution 0.0246 mol/l (0.0246 N) in aqueous solution
Supplier: Hach
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
2409232
HACH2409232EA 26.3 EUR
HACH2409232
Sodium thiosulphate 0.0246 mol/l (0.0246 N) in aqueous solution 0.0246 mol/l (0.0246 N) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844