BOD bottles
Supplier: Hach
62100 62106 62124
HACH62100EA 37.4 EUR
HACH62100 HACH62106 HACH62124
BOD bottles
Bottles BOD Bottles
These are standard bottles, made of borosilicate glass, for BOD analaysis.
  • Flared lip and ground-glass stopper to prevent air entrapment and for water seal
  • Serialised bottles have silk-screened, sequential numbers and include a bar code
  • Frosted marking area
Delivery information: Stoppers are included.