Ion selective electrodes, Fluoride, IntelliCAL™
Supplier: Hach
ISEF12103 ISEF12101 ISEF121AP
HACHISEF12103EA 1460 EUR
HACHISEF12103 HACHISEF12101 HACHISEF121AP
Ion selective electrodes, Fluoride, IntelliCAL™
Electrodes ISE Electrodes
IntelliCAL™ ISEF121 is a digital combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temperature sensor. The solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Requires virtually no maintenance
  • Fast, stable, and accurate results
  • Ultimate traceability in measurement history
  • Can be moved between meters without the need to re-calibrate or re-enter measurement settings
The probe is availableMore Product Information