Shakers, purging method
Supplier: Hach
LQV1489900001
HACHLQV1489900001EA 1150 EUR
HACHLQV1489900001
Shakers, purging method
Shakers Rotating/Rocking Shakers
Shaker which is used for purging of inorganic carbon (TIC). It has an automatic shutoff system. Accommodates eight cuvettes simultaneously.