Test Sieve with wooden frame, 300 x 300 x 80 mm Metal Wire Cloth made from Stainless Steel, Apertures according to DIN ISO 3310-1: 1 * 1 items
Supplier: NEXOPART
Test Sieve with wooden frame, 300 x 300 x 80 mm Metal Wire Cloth made from Stainless Steel, Apertures according to DIN ISO 3310-1: 1 * 1 items