Wire baskets, square
Supplier: GLASWARENFABRIK KARL HECHT
41075016 41075020
HECH41075016EA 110 EUR
HECH41075016 HECH41075020
Wire baskets, square
Baskets Wire Baskets
These square baskets are made of stainless steel. Designed for transporting, storing, cleaning and sterilising laboratory utensils.
  • Excellent temperature resistance
  • Good chemical resistance
  • Reinforced edges