Laboratory/media bottles, narrow neck, round, borosilicate glass, with screw cap
Supplier: GLASWARENFABRIK KARL HECHT
41330112 41330118 41330124 41330130 41330148 41330142 41330136
HECH41330112EA 4.7 EUR
HECH41330112 HECH41330118 HECH41330124 HECH41330130 HECH41330148 HECH41330142 HECH41330136
Laboratory/media bottles, narrow neck, round, borosilicate glass, with screw cap
Bottles Glass Bottles
Borosilicate glass 3,3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round, with graduations, DIN GL 45 thread, pouring ring and blue PP screw cap.