Sleeves for ground joints
Supplier: GLASWARENFABRIK KARL HECHT
42388010 42388019 42388024 42388029
HECH42388010EA 20 EUR
HECH42388010 HECH42388019 HECH42388024 HECH42388029
Sleeves for ground joints
NS Joint Sleeves and Connectors Joint Sleeves
Disposable sleeves for ground joints and stoppers, prevent the ground joints from sticking together.
  • Made of PTFE
  • Self-lubricating
  • Outstanding chemical resistance at temperatures