Separating funnels, conical
Supplier: GLASWARENFABRIK KARL HECHT
42951030 42951024 42951060 42950024 42950030 42950048 42950060 42950066 42950072
HECH42951030EA 44.4 EUR
HECH42951030 HECH42951024 HECH42951060 HECH42950024 HECH42950030 HECH42950048 HECH42950060 HECH42950066 HECH42950072
Separating funnels, conical
Funnels Separating Funnels
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, with solid glass or interchangeable PTFE key and plastic stopper.
  • Large labeling field
  • Excllent temperature resistance
  • Excllent chemical resistance
Certifikace: DIN ISO4800