Magnetic stirring bar, set
Supplier: Heidolph Instruments GmbH & Co.KG
509-58300-00
HEID509-58300-00EA 179 EUR
HEID509-58300-00
Magnetic stirring bar, set
Stirrers Magnetic Stirring Bars
Set of different shaped PTFE covered magentic stirring bars.
  • Very good chemical resistance
  • Temperature resistant, suitable for high and low temperature applications
Set includes 2× each of stirrers in a plastic box, stirrers have the following shapes and dimensions, L (mm): Cross shape 10 and 16,5 mm, elliptical shape 25 mm, oval 40 and 50 mm.