Bottle racks
Supplier: MERCK LIFE SCIENCE
1465250001 1465260001
HEIP1465250001EA 241 EUR
HEIP1465250001 HEIP1465260001
Bottle racks
Racks Bottle Racks
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
  • Autoclavable
  • Easy to clean
  • Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.