Silicone tubing on spool, HelixMark® Standard Tubing
Supplier: Freudenberg Medical Europe GmbH
60-375-03
HELI60-375-03EA 1230 EUR
HELI60-375-03
Silicone tubing on spool, HelixMark® Standard Tubing
Tubing Transfer Tubing
HelixMark® Standard silicone tubing meets the needs of a broad spectrum of medical and laboratory applications and are universally applicable for the transfer of fluids under standard requirements.
  • Fully autoclavable and sterilizable
  • Utilisation of platinum-cured silicone
  • Seamless batch traceability
Ideal for use in transfer operations as well as other applications requiring non-leachable material.
Certifikace: HelixMark® products are manufactured in clean rooms in compliance with the requirements of the medtech and pharmaceutical industry.