Magnetic Stir Bar, PTFE-Coated
Supplier: HELLMA
332-300-VE10
HELL332-300-VE10EA 49.3 EUR
HELL332-300-VE10
Magnetic Stir Bar, PTFE-Coated
Stirrers Magnetic Stirring Bars
For use with cell types: 109.000, 109.000F, 109.004, 109.004F, 119.000, 119.000F, 119.004, and 119.004F.
  • PTFE-coated
  • Length: 6 to 7 mm
  • 3 to 4 mm Ø